Jod standardised solution 0.01 N vandig løsning
Leverandør: Thermo Fisher Scientific
35636.K2
35636.K2EA 905 NOK
35636.K2
Jod standardised solution 0.01 N vandig løsning
Jod
CAS No.: 7553-56-2 Molecular Formula: I2 Formula Weight: 253.81 Physical Form: Liquid MDL No.: MFCD00011355
Formel: I₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011355
CAS-nummer: 7553-56-2
EINECS: 231-442-4
FN: 3264
ADR: 8,II