Natriumhydroksid 20% (w/v) in aqueous solution

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: Caustic soda

35637.AP
35637.APEA 465 NOK
35637.AP
Natriumhydroksid 20% (w/v) in aqueous solution
Natriumhydroksid

CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid UN No.: 1824 MDL No.: MFCD00003548

Formel: NaOH
MW: 40 g/mol
Kokepunkt: 119…135 °C (1013 hPa)
Smeltepunkt: 9…10 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1824
ADR: 8 II
Merck Indeks: 14,08627

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR