Svovelsyre standardised solution 0.1 N vandig løsning

Leverandør: Thermo Fisher Scientific

35651.K2 35651.M4
35651.K2EA 545 NOK
35651.K2 35651.M4
Svovelsyre standardised solution 0.1 N vandig løsning
Svovelsyre

Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid MDL No.: MFCD00064589

Formel: H₂SO₄
Tetthet: 1,01 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064589
CAS-nummer: 7664-93-9
EINECS: 231-639-5
FN: 2796
ADR: 8,II

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR