Glysin, Millipore®

Leverandør: Merck Millipore Calbiochem

357002-1
357002-1EA 2600 NOK
357002-1
Glysin, Millipore®
Glysin
Formel: NH₂CH₂COOH
Kokepunkt: 181 °C (1013 hPa)
Smeltepunkt: 83 °C
Tetthet: 1,64 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008131
CAS-nummer: 56-40-6
EINECS: 200-272-2
Merck Indeks: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR