Cobalt(II) fluoride, ultra dry ≥99.99% (metals basis)

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: Cobalt difluoride, Cobaltous fluoride

Ampuled under argon

35740.04 35740.09
35740.04EA 1175 NOK
35740.04 35740.09
Cobalt(II) fluoride, ultra dry ≥99.99% (metals basis)
Cobalt(II) fluoride

Purity: 99.99% CAS No.: 10026-17-2 Molecular Formula: CoF2 Formula Weight: 96.93 Physical Form: Powder, Ampouled under argon Density: 4.46 Melting Point: 1200° Boiling Point: 1400° Merck Reference: 14,2440 UN No.: 2923 MDL No.: MFCD00010941

Formel: CoF₂
MW: 96.93 g/mol
Kokepunkt: 1400 °C
Smeltepunkt: 1200
Tetthet: 4.46
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010941
CAS-nummer: 10026-17-2
EINECS: 233-061-9
FN: 2923
ADR: 8 III
Merck Indeks: 14,2440

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR