Iridium 1000 mg/L Ir in 20% saltsyre (fra IrCl₃) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

35751.AD 35751.AE 35751.AP
35751.ADEA 1700 NOK
35751.AD 35751.AE 35751.AP
Iridium 1000 mg/L Ir in 20% saltsyre (fra IrCl₃) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Iridium Standards

Molecular Formula: IrCl₃ in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011062


Sertifiseringer: Traceable to NIST

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00011062

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR