Iridium 10000 mg/L Ir in 20% saltsyre (fra IrCl₃) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

35752.AD 35752.AE 35752.AP
35752.ADEA 6200 NOK
35752.AD 35752.AE 35752.AP
Iridium 10000 mg/L Ir in 20% saltsyre (fra IrCl₃) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Iridium Standards

Molecular Formula: IrCl₃ in 20% HCl
Physical Form: Liquid
Odour: Characteristic
UN No.: 1789
MDL No.: MFCD00011062


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR