Europium 1000 mg/L Eu in 5% salpetersyre (fra Eu₂O₃) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

35753.AD 35753.AE 35753.AP
35753.ADEA 995 NOK
35753.AD 35753.AE 35753.AP
Europium 1000 mg/L Eu in 5% salpetersyre (fra Eu₂O₃) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Europium Standards

Molecular Formula: Eu₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010992


Sertifiseringer: Traceable to NIST

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00010992

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR