Lutetium 10.000 mg/l Lu in 5% salpetersyre (fra Lu2O3) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

35757.AEEA 9075 NOK
35757.AE
Lutetium 10.000 mg/l Lu in 5% salpetersyre (fra Lu2O3) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Lutetium Standards

Molecular Formula: Lu2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011098


Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR