Supelco® PESTANAL® Organic Reference Standard, Phosalone

Leverandør: Merck

36194-100MG
36194-100MGEA 1050 NOK
36194-100MG
Supelco® PESTANAL® Organic Reference Standard, Phosalone
Standarder Organiske standarder

  • GHS signal: GHS06, GHS09


NB: P261 - P273 - P280 - P301 + P310 - P302 + P352 + P312 - P304 + P340 + P312

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66334112dda25db306e31c26d721cc5c3fe0d95f&vid=66334112dda25db306e31c26d721cc5c3fe0d95f&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 2310-17-0
MDL No. MFCD00055499
Formel C₁₂H₁₅ClNO₄PS₂
Molekylvekt 367,81
Hardhetsklasse beskrivelse H301 + H311 - H317 - H332 - H410
Flammepunkt 100 °C - closed cup
Fraktforhold STD
Lagringsforhold 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR