Natriumklorid ≥99.0% (by argentometric titration), krystaller, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Warning

3624-20EA 6900 NOK
3624-20 3624-01 3624-05 3624-06 3624-07 3624-60
Natriumklorid ≥99.0% (by argentometric titration), krystaller, BAKER ANALYZED® ACS, J.T.Baker®
Natriumklorid
Formel: NaCl
Kokepunkt: 1413 °C (1013 hPa)
Smeltepunkt: 801 °C
Tetthet: 2,16 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003477
CAS-nummer: 7647-14-5
Merck Indeks: 14,08599

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements ≥99.0%
For Laboratory,Research,or Manufacturing Use 5.0 - 9.0
Assay (NaCl) (by Ag titrn) ≥99.0 %
pH of 5% Solution at 25°C 5.0 - 9.0
Insoluble Matter ≤0.005 %
Iodide (I) ≤0.002 %
Bromide (Br) ≤0.01 %
Chlorate and Nitrate (as NO₃) ≤0.003 %
ACS - Phosphate (PO₄) ≤5 ppm
Sulfate (SO₄) ≤0.004 %
Barium (Ba) Passes Test
ACS - Heavy Metals (as Pb) ≤5 ppm
Iron (Fe) ≤2 ppm
Calcium (Ca) ≤0.002 %
Magnesium (Mg) ≤0.001 %
Potassium (K) ≤0.005 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR