Antimon(III)klorid ≥99.0% ACS

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: Antimony trichloride

36281.30 36281.A1 36281.18
36281.30EA 3200 NOK
36281.30 36281.A1 36281.18
Antimon(III)klorid ≥99.0% ACS
Antimon(III)klorid

Purity: 99.0% min CAS No.: 10025-91-9 Molecular Formula: SbCl3 Formula Weight: 228.11 Physical Form: Crystalline Density: 3.14 Melting Point: 73.4° Boiling Point: 223.5° Solubility: Soluble in water with hydrolysis to SbOCl. Soluble in HCl solution, alcohol, benzene, carbon disulfide, dioxane, chloroform, ether, acetone, carbon tetrachloride. Insoluble in organic bases Merck Reference: 14,707 UN No.: 1733 MDL No.: MFCD00011212


Stripping tin coating from steel surfaces, in organic synthesis, as catalyst, in making other antimony salts, in chemical microscopy for drug identification

Formel: SbCl₃
MW: 228.12 g/mol
Kokepunkt: 223.5 °C
Smeltepunkt: 73.4 °C
Tetthet: 3.14
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011212
CAS-nummer: 10025-91-9
EINECS: 233-047-2
FN: 1733
ADR: 8 II
Merck Indeks: 14,707

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR