tri-Natriumcitrat dihydrat 99.0-101.0% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor

3646-01EA 885 NOK
3646-01 3646-07 3646-R
tri-Natriumcitrat dihydrat 99.0-101.0% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
tri-Natriumcitrat dihydrat

Granular. Lot analysis on label.


  • Lot analysis on label

Formel: Na₃C₆H₅O₇·2H₂O
Smeltepunkt: 150 °C
Tetthet: 1,70 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150031
CAS-nummer: 6132-04-3
EINECS: 200-675-3
Merck Indeks: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements 99.0 - 101.0 %
For Laboratory,Research,or Manufacturing Use 99.0 - 101.0 %
ACS - Assay (C₆H₅Na₃O₇·2H₂O) (by non-aqueous titration) 99.0 - 101.0 %
ACS - pH of 5% Solution at 25°C 7.0 - 9.0
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤0.003 %
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Ammonia (as NH₃) ≤0.003 %
ACS - Calcium (Ca) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR