Supelco® PESTANAL® Organic Reference Standard, Clopyralid (2-Hydroxyethyl)Ammonium

Leverandør: Merck

36529-250MG
36529-250MGEA 595 NOK
36529-250MG
Supelco® PESTANAL® Organic Reference Standard, Clopyralid (2-Hydroxyethyl)Ammonium
Standarder Organiske standarder

  • GHS signal: GHS05


NB: P280 - P305 + P351 + P338

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f6404a7a6aebe9208ebc3ad32cb4ee7548df036&vid=4f6404a7a6aebe9208ebc3ad32cb4ee7548df036&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) 3,6-Dichloropyridine-2-carboxylic acid ethanolammonium salt
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 57754-85-5
MDL No. MFCD00777778
Molekylvekt 253,08
Hardhetsklasse beskrivelse H318
Flammepunkt >100 °C
Fraktforhold STD
Lagringsforhold 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR