Supelco® PESTANAL® Organic Reference Standard, Chlorfenvinphos

Leverandør: Merck

36551-250MG
36551-250MGEA 750 NOK
36551-250MG
Supelco® PESTANAL® Organic Reference Standard, Chlorfenvinphos
Standarder Organiske standarder

  • GHS signal: GHS06, GHS09


NB: P264 - P270 - P273 - P280 - P264 - P273 - P280 - P301 + P310 - P302 + P352 + P312 - P301 + P310 - P302 + P352 + P312 - P361 + P364

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f75a0fb8a17be2440bde5326aa2297042fe9723&vid=0f75a0fb8a17be2440bde5326aa2297042fe9723&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 470-90-6
MDL No. MFCD00041818
Molekylvekt 359,57
Hardhetsklasse beskrivelse H300 - H311 - H410 - H300 - H311 - H410
Fraktforhold STD
Lagringsforhold 2...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR