Supelco® TraceCERT® Organic Reference Standard, Haloxyfop-P

Leverandør: Merck

36824-25MG
36824-25MGEA 840 NOK
36824-25MG
Supelco® TraceCERT® Organic Reference Standard, Haloxyfop-P
Standarder Organiske standarder

Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.


NB: P264 - P270 - P273 - P301 + P310 - P391 - P405

Bestill nå

SPESIFIKASJONER

Synonym(s) (R)-2-[4-(3-Chloro-5-(trifluoromethyl)-2-pyridyloxy)phenoxy]propionic acid
Environmentally Preferable
M-Clarity quality level MQ300
CAS-nr 95977-29-0
MDL No. MFCD20036282
Molekylvekt 361,70
Hardhetsklasse beskrivelse H301 - H410
Fraktforhold WICE
Lagringsforhold 2...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR