Guanidiniumtiocyanat, Millipore®

Leverandør: Merck Millipore Calbiochem
Warning

Synonymer: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

368975-500
368975-500EA 8825 NOK
368975-500
Guanidiniumtiocyanat, Millipore®
Guanidiniumtiocyanat
Formel: CH₅N₃·CHNS
MW: 118.16 g/mol
Smeltepunkt: 118
Tetthet: 1.29 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013027
CAS-nummer: 593-84-0
EINECS: 209-812-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR