Tetrahydrofuran ≥99.8% stabilised, AR®, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger


Stabilisering: stabilisert med BHT/Ionol 250 ppm

3705-45EA 10650 NOK
3705-45 3705-25
Tetrahydrofuran ≥99.8% stabilised, AR®, Macron Fine Chemicals™
Tetrahydrofuran

NB: Preserve in tight and cool conditions.

Formel: C₄H₈O
Kokepunkt: 65 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005356
CAS-nummer: 109-99-9
EINECS: 203-726-8
FN: 2056
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Assay (GC) ≥99,8
Water (KF) ≤0,03%
Titrable Acid (meq/g) ≤0,0005
Residue after Evaporation ≤0,03%
Stabiliser (BHT) ~250 ppm
For Laboratory, Research or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR