Guanosine 5'-triphosphate disodium salt, Millipore®

Leverandør: Merck Millipore Calbiochem

371701-250
371701-250EA 6700 NOK
371701-250
Guanosine 5'-triphosphate disodium salt, Millipore®
Guanosine 5'-triphosphate disodium salt
Formel: C₁₀H₁₄N₅Na₂O₁₄P₃
MDL nummer: MFCD03410297
CAS-nummer: 56001-37-7
EINECS: 259-940-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR