GW 9508 98 HPLC_ASSAY_METHOD, Calbiochem®, Sigma-Aldrich®

Leverandør: Merck Millipore Calbiochem

Synonymer: GPR40 Agonist, GW9508, 4-[[(3-Phenoxyphenyl)methyl]amino]benzene propanoic acid, GW-9508, 3-(4-((3-Phenoxybenzyl)amino)phenyl)propanoic acid, 3-(4-(((3-(Phenoxy)phenyl)methyl)amino)phenyl)propanoic acid, Free Fatty Acid Receptor 1 Agonist I, FFA1 Agonist I

MQ100

371706-10
371706-10EA 4250 NOK
371706-10
GW 9508 98 HPLC_ASSAY_METHOD, Calbiochem®, Sigma-Aldrich®
GW 9508

The GPR40 Agonist, also referenced under CAS 885101-89-3, controls the biological activity of GPR40. This small molecule/inhibitor is primarily used for Cell Signaling applications.


General description
A cell-permeable aminophenylpropanoate that acts as a potent and selective agonist for the G-protein coupled receptor (GPCR) GPR40 (EC50 ~50 nM) with much reduced activity towards family members GPR120 (EC50 ~3.5 µM), GPR41/GPR43 (EC50 >50 µM), as well as a panel of eight other fatty acid and prostaglandin receptors. Shown to induces GPR40-dependent Ca2+ mobilization in HEK-293 cells and potentiate glucose-stimulated insulin secretion in MIN6 cells. Reported to exhibit an in vivo half-life of ≥5.3 hr. in rat (p.o. or i.v.) with a bioavailability of 65% and a clearance of 24.9 ml/min/kg.
A cell-permeable aminophenylpropanoate that acts as a potent and selective agonist for the G-protein coupled receptor (GPCR) GRP40 (EC50 ~50 nM) with much reduced activity towards family members GRP120 (EC50 ~3.5 µM), GRP41/GRP43 (EC50 >50 µM), as well as a panel of eight other fatty acid and prostaglandin receptors. Shown to induces GPR40-dependent Ca2+ mobilization in HEK-293 cells and potentiate glucose-stimulated insulin secretion in MIN6 cells. Reported to exhibit an in vivo half-life of ≥5.3 hr. in rat (p.o. or i.v.) with a bioavailability of 65% and a clearance of 24.9 ml/min/kg.


Forpakning: Packaged under inert gas

Formel: C₂₂H₂₁NO₃
MW: 347.414 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD09753282
CAS-nummer: 885101-89-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR