Basic blue 3 25% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: BLUE DE LYON, CI BASIC BLUE 3, BasicblueBG

M-Clarity™ quality level = MQ100

378011-100G
378011-100GEA 1890 NOK
378011-100G
Basic blue 3 25% (dye content), Sigma-Aldrich®
Basic blue 3

Basic blue 3 (BB3) can be removed from aqueous solutions with the help of activated carbon prepared from rubber seed coat.[1] Basic blue 3 (C.I. 51104) is a cationic dye. It is generally positively charged in an aqueous solution. Basic blue 3 has a brilliant blue shade and thus has been used as a colourant in dyeing acrylic fabrics.

Formel: C₂₀H₂₆N₃OCl
MW: 359.9 g/mol
Smeltepunkt: 205 °C (dec., lit.)
Lagringstemperatur: Room Temperature
MDL nummer: MFCD00011928
CAS-nummer: 33203-82-6
EINECS: 251-403-5

Bestill nå

Spesifikasjon Testresultater

Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Carbon Specification: Report Results
Nitrogen Specification: Report Results
Wavelength, c = 0.01 g/L; Water 651 - 657 nm
Extinction Coefficient ≥12000
Wavelength 257 - 263 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR