Natriumdihydrogenortofosfat monohydrat 98.0-103.0% (by anhydrous basis), krystaller USP, FCC, J.T.Baker®

Leverandør: Avantor

3820-01EA 1550 NOK
3820-01 3820-07
Natriumdihydrogenortofosfat monohydrat 98.0-103.0% (by anhydrous basis), krystaller USP, FCC, J.T.Baker®
Natriumdihydrogenortofosfat monohydrat

  • Food GMP manufactured product

Formel: NaH₂PO₄·H₂O
Smeltepunkt: 100 °C
Tetthet: 2,04 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149208
CAS-nummer: 10049-21-5
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Indeks: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Meets U.S.P Requirements 98.0 - 103.0%
GMP Manufactured Product Passes Test
Meets F.C.C. Requirements ≤3 ppm
Food GMP Manufactured Product ≤0.014%
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Food Chemical Passes Test
Bulk Pharmaceutical Chemical ≤0.2%
USP - Assay (anhydrous basis) 98.0 - 103.0 %
USP - Aluminum, Calcium, and Related Elements Passes Test
USP - Arsenic (As) ≤3 ppm
USP - Chloride (Cl) ≤0.014 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Substances ≤0.2 %
USP - pH (1 in 20) 4.1 - 4.5
USP - Sulfate (SO₄) ≤0.15 %
USP - Water (H₂O) 10.0 - 15.0 %
FCC - Assay (anhydrous basis) 98.0 - 103.0 %
FCC - Arsenic (As) ≤3 ppm
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Fluoride (F) ≤0.005 %
FCC - Insoluble Substances ≤0.2 %
FCC - Lead (Pb) ≤4 mg/kg
FCC - Loss on Drying 10.0 - 15.0 %
Appearance (White crystals or granules) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR