Dinatriumhydrogenortofosfat 98.0-100.5% (dried basis) USP, J.T.Baker®

Leverandør: Avantor
Warning


Management of Change (MOC) category = HR

3827-01EA 1250 NOK
3827-01 3827-07 3827-19
Dinatriumhydrogenortofosfat 98.0-100.5% (dried basis) USP, J.T.Baker®
Dinatriumhydrogenortofosfat

  • Food GMP manufactured product

Formel: Na₂HPO₄
Smeltepunkt: 250 °C
Tetthet: 2,07 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003496
CAS-nummer: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Indeks: 12,08805

Bestill nå

Spesifikasjon Testresultater

GMP Manufactured Product 98.0 - 100.5%
Meets U.S.P Requirements ≤3 ppm
CAUTION: For Manufacturing, processing or repackaging ≤0.06%
Bulk Pharmaceutical Chemical Passes Test
USP - Assay (Na₂HPO₄) (dried basis) 98.0 - 100.5 %
USP - Arsenic (As) ≤ 3 ppm
USP - Chloride (Cl) ≤ 0.06 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Matter ≤ 0.2 %
USP - Loss on Drying at 130°C ≤ 5.0 %
USP - Sulfate (SO₄) ≤ 0.2 %
(Suitable for Buffer Solutions) ≤0.005%
No Class 1,2,3 or other solvents are used or produced in the ≤0.2%
manufacturing or purification of the product. ≤4 mg/kg
Elemental Impurities (USP 232, EP 5.20) - Information on elemental ≤5.0%
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR