Palladium 5% on activated carbon powder wetted with ca. 50% water for use as a catalyst, unreduced

Leverandør: Thermo Fisher Scientific

38301.22
38301.22EA 0 NOK
38301.22
Palladium 5% on activated carbon powder wetted with ca. 50% water for use as a catalyst, unreduced
Palladium

Physical Form: 900-1100 m²/g Odor: Odorless


Bestillingsinformasjon: Sold on a dry weight basis. Unit weight excludes water weight

Formel: Pd
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011167
CAS-nummer: 7440-05-3

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR