2,3-Dimerkapto-1-propanol ≥98.0% (by GC) for complexometry, for spectrophotometric determination of Cu, Fluka™

Leverandør: Honeywell Chemicals
Danger

38520-25ML
38520-25MLEA 8550 NOK
38520-25ML
2,3-Dimerkapto-1-propanol ≥98.0% (by GC) for complexometry, for spectrophotometric determination of Cu, Fluka™
2,3-Dimerkapto-1-propanol
Formel: C₃H₈OS₂
Kokepunkt: 120 °C (20 hPa)
Smeltepunkt: 77 °C
Tetthet: 1,24 g/cm³ (25 °C)
Flash punkt: 112 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004864
CAS-nummer: 59-52-9
EINECS: 200-433-7
FN: 2810
ADR: 6.1,III
Merck Indeks: 13,03234
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=745c14e3244d75cab02b536ae9d18ec21829aeba&vid=345ebd39845f4c372f03844bee87b4d8c249ec2b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Refractive Index n20/D 1.574
Assay (GC) Min. 98.0 Fl.%
IR spectrum complying
Refractive index (n 20/D) 1.573 - 1.575


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR