Giemsa's stain solution, J.T.Baker®

Leverandør: Avantor
Danger

Synonymer: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

3856.1000EA 580 NOK
3856.1000
Giemsa's stain solution, J.T.Baker®
Giemsa's stain

GIEMSA's solution Azur-Eosine-methylene blue solution for microscopy (contains methanol).

MDL nummer: MFCD00081642
CAS-nummer: 51811-82-6
FN: 1992
ADR: 3 II
IVDLogo

Bestill nå

Spesifikasjon Testresultater

Performance Test Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR