Naphthol AS-BI phosphate solution_SC 12,5 mg/mL DMF, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Danger

Synonymer: 7-Bromo-3-hydroxy-2-naphthoic-o-anisidide phosphate, Naphthol AS-BI Phosphoric acid solution, Naphthol AS-BI-Phosphate, 6-Bromo-3-((2-methoxyphenyl)carbamoyl)naphthalen-2-yl dihydrogen phosphate

MQ500

3864-40ML
3864-40MLEA 1490 NOK
3864-40ML
Naphthol AS-BI phosphate solution_SC 12,5 mg/mL DMF, Sigma-Aldrich®
Naphthol AS-BI phosphate

Naphthol AS-BI Phosphoric acid solution foruse in Sigma Acid Phosphatase Procedure No. 386.


  • Suitable for haematology
  • Suitable for histology

Formel: C₁₈H₁₅BrNO₆P
MW: 452.1926 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00004058
CAS-nummer: 1919-91-1
FN: 2265
ADR: 3 III
IVDLogo

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR