Fluorsyre 47 - 51%, Environmental Grade Plus

Leverandør: Thermo Fisher Scientific
Danger

38746.AP
38746.APEA 22800 NOK
38746.AP
Fluorsyre 47 - 51%, Environmental Grade Plus
Fluorsyre

CAS No.: 7664-39-3 Molecular Formula: HF Formula Weight: 20.01 Physical Form: Liquid, double sub-boiling quartz distillation Density: 1.16 Appearance: Colourless Odour: Acrid Boiling Point: 112° Merck Reference: 14,4790 UN No.: 1790 MDL No.: MFCD00011346

Formel: HF
Kokepunkt: 112 °C
Tetthet: 1,16
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011346
CAS-nummer: 7664-39-3
EINECS: 231-634-8
FN: 1790
ADR: 8,II
Merck Indeks: 14,4790

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR