Inkubator, mikrobiologisk, med eller uten vifte, BD- og BF-serien
Leverandør: Binder
9010-0073 9010-0187 9010-0241
390-6034EA 71250 NOK
390-6034 390-6146 390-0009
Inkubator, mikrobiologisk, med eller uten vifte, BD- og BF-serien
Inkubatorer Inkubatorer, standard
Two versions of these incubators with electronically controlled APT.line™ preheating chamber are available depending on the application: The BD series with natural convection for all standard applications and the BF series with forced convection for applications with high load density.
  • Additional safety is provided with disinfection at 100 °C and easy operation
  • DS controller with integrated timer 0 to 99 hours (BD series), MS controller with several timer functions such as 'Delayed OFF' and 'Delayed ON' (BF series)
  • High degree of temperature accuracy and