CO₂ incubator, Midi 40
Leverandør: Thermo Scientific
3404
390-0456EA 74700 NOK
390-0456
CO₂ incubator, Midi 40
Inkubatorer CO₂-inkubatorer
The Midi 40 CO₂ incubator is designed specifically for people who require a compact culturing workspace to handle small workloads, perform personal use applications, or have limited workspace. The unit has the same performance and capabilities as full sized incubators and is a good solution to avoid shared-use environments that can present potential contamination risk through unnecessary sample handling and repeated door openings.
  • Cost-effective dedicated incubator space which prevents cross contamination
  • Removable water reservoir provides up to 95% RH