Constant climate chambers with Advanced Peltier Technology, HPPeco series
Leverandør: MEMMERT
HPP1060ECO HPP110ECO HPP1400ECO HPP2200ECO HPP260ECO HPP410ECO HPP750ECO
390-1516EA 0 NOK
390-1516 390-1517 390-1518 390-1521 390-1522 390-1523 390-1524
Constant climate chambers with Advanced Peltier Technology, HPPeco series
Testkammer Klimakammer
Maximum energy effiency in continous operation - these powerful constant climate chambers work with the new Advanced Peltier Technology. These devices with TwinDISPLAY are ideal for stability studies according to ICH guidelines, stability tests for cosmetics and foods as well as environmental and material testing.
  • Maximum energy efficiency compared to climate chambers with compressor
  • Completely homogenous temperature and humidity distribution
  • Reliable and fail-safe
  • Refrigerant-free and low maintenance
These powerful constant climate chambers areMore Product Information