CO₂-inkubator med styreskinner for WHEATON® rulleapparat, WHEATON®
Leverandør: DWK Life Sciences
WI057608-C-E
390-1588EA 0 NOK
390-1588
CO₂-inkubator med styreskinner for WHEATON® rulleapparat, WHEATON®
Inkubatorer CO₂-inkubatorer
WHEATON® CO₂ Incubators with an IR CO₂ sensor are ideal for everyday cell culture work and designed to be used with the WHEATON® Roller Apparatus.
  • Guide tracks in base for ease of roller apparatus entry
  • User configurable shelving - up to 30 shelves
  • Designed for temperature uniformity
  • Recovery time after 30 seconds door opening at 37 °C is less than 5 minutes
  • CO₂ recovery time after 30 seconds door opening at 5 Vol.% CO₂ is less than 5 minutes
  • Large viewing window in the door (with an option for dual windows)
  • Infrared sensor technology to maintain 0 to 20%
More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Temperatursvingning (tid) ±0.1 @ 37 °C
Temperaturvariasjon (romlig) ±0,5 °C @ 37 °C
Kapasitet 1118 L
Konveksjontypen tvungen konveksjon
Maks. belastning per hylle 34 kg
Max. chamber workload 204 kg
Nominelt strømforbruk 9 A
Temperaturområde Ambient +8…60 °C
Vekt 260,4 kg