Heratherm™ Environmental Chambers
Leverandør: Thermo Fisher Scientific
ESS750 ESG750
390-1712EA 0 NOK
390-1712 390-1713
Heratherm™ Environmental Chambers
Testkammer Klimakammer
Heratherm™ environmental chambers offer exceptional environmental control in a hassle-free chamber that is easy to program and use.
  • Reduce your environmental footprint while achieving outstanding results with Heratherm environmental chambers decreased water and energy consumption and Peltier technology
  • Water-efficient humidification system with integrated water tank and optional clean water connection
  • Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors
  • Vertical low-energy LED lighting