Små inkubatorer, B28
Leverandør: Binder
9010-0002
390-6032EA 17175 NOK
390-6032
Små inkubatorer, B28
Inkubatorer Inkubatorer, bordmodell
A compact device with hydraulic-mechanical control for precise, reliable incubation conditions. Optimum temperature range at 37 °C. Unit has galvanised sheet steel housing, powder coated throughout to afford maximum protection against corrosion, hydraulic-mechanical thermostat with analogue dial thermometer.
  • Available with or without independent temperature safety regulator TC Class 1- in the event of all other controllers failing, mechanical temperature limiter Class 1 stops the heating at ~10 °C above the maximum temperature of the incubator, saves the