Boats, glassy carbon
Leverandør: Thermo Fisher Scientific
39026.KT 39027.KT 39028.KT
39026.KTEA 2975 NOK
39026.KT 39027.KT 39028.KT
Boats, glassy carbon
Skip Forbrenningsskip
Combustion boats, glassy carbon.
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Resistive to abrasive wear
  • Good electrical conductivity
Boats for high-temperature differential thermal/thermal gravimetric analyses. Good resistance to thermal shock allows rapid heating and cooling times.