Urine collection bottles, for women
Leverandør: GLASWARENFABRIK KARL HECHT
44104010
391-0024EA 170 NOK
391-0024
Urine collection bottles, for women
Diagnostiske systemer Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Urinals made of PE for women.
  • Natural coloured
  • Approximate scale, with cover
  • Heat resistant up to 90 °C