Flasker med rulleflens, liten åpning, DURAN®
Leverandør: DWK Life Sciences
214612403
391-0025EA 305 NOK
391-0025
Flasker med rulleflens, liten åpning, DURAN®
Flasker Cellekulturflasker
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For sampling and cultivation.