Septum caps, Nalgene®
Leverandør: Thermo Fisher Scientific
DS2168-0384
391-0073EA 870 NOK
391-0073
Septum caps, Nalgene®
Lokk Flaskelokk
Polypropylene closure with thermoplastic elastomer septum.
  • Use with 18-gauge or smaller needles
For Nalgene bottles with 38-430 threaded mouth.