Temperature indicating tubes
Leverandør: KOEHLER TECHNISCHE PRODUKTEN
K-1253 K-1252 K-1250
391-0084EA 1950 NOK
391-0084 391-0085 KOEHK-1250
Temperature indicating tubes
Temperatur indikatorstrips
Temperature indicating tubes are used for autoclaves and hot air sterilisers.
Sterilisation is detected by the colour change of the control tube from red to green. The tubes should be placed as close as possible next to or in the load.