Sealing tape with dispenser, Nunc™
Leverandør: Thermo Fisher Scientific
236707
391-0113EA 8900 NOK
391-0113
Sealing tape with dispenser, Nunc™
Forseglinger Filmer og folier til mikroplateforsegling
Polyester tape with acrylic adhesive. Easy peel, easy seal general-purpose tape with a faster application time than normal tape in loose sheets.
  • Temperature stable from −20 to +120 °C
  • Non-pierceable by pipette
Colour: clear
External L×W: 121×78 mm (plus handle 9×78 mm)