Roux-kulturkolber, DURAN®
Leverandør: DWK Life Sciences
215415801 215715804
391-0360EA 1175 NOK
391-0360 391-0170
Roux-kulturkolber, DURAN®
Kolber Cellekulturkolber
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Round neck (391-0360)
  • Conical neck, eccentric (391-0170)
Typical application: preparation of cultures in nutrient media.