VWR®, Medieflasker med kork
Leverandør: VWR Collection
391-0283EA 1950 NOK
391-0283 391-0284 391-0286 391-0299
VWR®, Medieflasker med kork
Flasker Cellekulturflasker
PS, PC eller PETG, med PP-skrukork, steril.
  • Sloped design improves bottle stability when placed on a work surface
  • Narrow bottle shoulder, combined with moulded finger grips, improves the ability to securely grip the bottle with gloved hands
  • Easy Grasp bottle cap facilitates uncapping, tightening and bottle stacking, as well as providing a convenient gripping surface while transporting
  • Polystyrene (PS) bottles are supplied with a white cap
  • Polycarbonate (PC) bottles are supplied with a blue cap
  • PETG bottles are supplied with a green cap
VWR® CollectionMore Product Information