Fernbach-kulturkolber, DURAN®
Leverandør: DWK Life Sciences
217746209
391-0310EA 1675 NOK
391-0310
Fernbach-kulturkolber, DURAN®
Kolber Cellekulturkolber
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Conical shape, straight neck for metal caps
With plain rim.
Typical application: preparation of cultures in nutrient media.