VWR®, Reagent Reservoir, 175 ml
Leverandør: VWR Collection
391-0625EA 470 NOK
391-0625
VWR®, Reagent Reservoir, 175 ml
Reagenstrau
175 ml graduated polypropylene reagent reservoir with lid. Unique V-shaped bottom reduces dead volume.
  • Easy open locking lid
  • Graduation marks at 25, 50, 75, 100, 125 and 150 ml
  • Excellent chemical resistance (PP)
  • Stackable
Can be autoclaved.