VWR® Adhesive Sealing Strips for 8-Strip PCR Tubes
Leverandør: VWR Collection
391-0638EA 845 NOK
391-0638
VWR® Adhesive Sealing Strips for 8-Strip PCR Tubes
Forseglinger Filmer og folier til mikroplateforsegling
Easier to apply and remove as compared to standard moulded PCR strip caps.
  • Ideal for PCR and storage
  • Alternative to strip-caps; no sore thumbs and fingers guaranteed
  • Seals a single 8-well row of a 96-well plate or a single PCR 8-tube strip
  • Certified DNase- and RNase-free
  • 2,0 mil transparent PE film
  • Recommended temperature range from −20 to +120 °C
8 strips are pre-cut on a continuous liner for ease of use.