VWR® Petri Dishes with Lid, Soda-lime Glass
Leverandør: VWR Collection
391-0652EA 470 NOK
391-0652 391-0660 391-0665 391-0668 391-0671 391-0673 391-0674 391-0707 391-0708
VWR® Petri Dishes with Lid, Soda-lime Glass
Dyrkningsplater Petriskåler
Glass Petri dishes are ideal for cultivating bacteria, fungi, and other microorganisms, as well as for cell cultures in research and diagnostics.
  • Durable, reusable, and easy to clean
  • A sustainable choice
  • Chemically inert glass eliminates unwanted reactions with cultures or samples
  • Transparent glass makes it easier to observe growth and development
Glass Petri dishes are durable, reusable, and they provide a sterile environment for various scientific applications.