VWR® Petri Dishes, Glass
Leverandør: VWR Collection
391-0698EA 1750 NOK
391-0698
VWR® Petri Dishes, Glass
Dyrkningsplater Petriskåler
Soda-lime glass, with lid. For use as disposables. These Petri dishes are a green alternative to disposable Petri dishes made from plastic.
  • Good chemical resistance
  • Heatable up to 60 °C
  • Not suitable for applications related to strong heat or rapid temperature changes
  • Ideal for bacterial culture production
NB: The indication of the diameter of the petri dishes is approximate.