Microplates, 1536-well
Leverandør: HIRSCHMANN
9240136
391-1972EA 0 NOK
391-1972
Microplates, 1536-well
Mikroplater Mikroplater med glassinnsats
Microtitration plate, made from PP, with fixed glass inserts.
  • High purity and chemically inert
  • 1536 wells
  • Translucent plastic cover included in standard scope of delivery
Plate dimensions: 124×81 mm