Stativ for petriskåler, Nalgene®
Leverandør: Thermo Fisher Scientific
5920-0060
391-2161EA 3050 NOK
391-2161
Stativ for petriskåler, Nalgene®
Stativer for rør Petriskåldispenser
Petri dish rack with PC frame and white PC posts.
  • Smooth, rounded corners
  • Finger-grip handle and rubber feet make carrying safe and secure
  • Rack can be decontaminated by heat sterilisation
For up to 54 plastic round dishes (100×15 mm) or 42 glass round dishes (100×15 mm). Simple assembly required.
W×D×H: 203×343×235 mm