Celleskraper
Leverandør: Greiner Bio-One
541070_100 541080_100
391-3010EA 5900 NOK
391-3010 391-3011
Celleskraper
Celleskrapere
Designed to facilitate the gentle mechanical removal of adherent cells from growth surfaces. Each scraper has a 18 mm pivoted (pivot angle 60°) scraper blade, which allows access to poorly accessible surfaces.
  • Optimised blade design for maximum cell harvest
  • Uniform contact with the growth surface, minimal mechanical strain and efficient cell harvest, even from poorly accessible surfaces
  • Minimises any accumulation of the cell suspension to the blade structure
Forpakning: pakket enkeltvis